Documenti fondamentali
BR36983
BRAND® volumetric flask BLAUBRAND® USP conformity certified
capacity 1000 mL, neck joint: ST/NS 24/29, glass stopper
Sinonimo/i:
BLAUBRAND®, USP, Volumetric flasks
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
glass stopper
Giunto
joint: ST/NS neck24/29
Caratteristiche
accuracy class A
Confezionamento
pack of 1
Produttore/marchio commerciale
BRAND 36983
Capacità
1000 mL
Descrizione generale
The BRAND® volumetric flask BLAUBRAND® USP conformity certified is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for a variety of laboratory applications. Each flask comes with a USP batch certificate, confirming its adherence to stringent quality and safety standards. This volumetric flask is an essential tool for precise analytical work, making it ideal for laboratories that require reliable and certified liquid handling solutions.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
BR36983-1EA4061831856095