BR36404
BRAND® BLAUBRAND® volumetric flask trapezoidal
capacity 10 mL, polypropylene stopper, joint: ST/NS 10/19
Sinonimo/i:
BLAUBRAND®, Volumetric flasks
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prodotti consigliati
Materiali
polypropylene stopper
transparent borosilicate glass 3.3 flask
Giunto
joint: ST/NS10/19
Caratteristiche
accuracy class A
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
BRAND 36404
Capacità
10 mL
Descrizione generale
Borosilicate 3.3. To DIN EN ISO 1042. Calibrated to contain (TC, In).
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
Note legali
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)