BR156648
BRAND® tray stackable
size 625 x 530 x 140 mm
Sinonimo/i:
Photographoc trays, Trays
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.51
Prodotti consigliati
Materiali
polypropylene
white
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 156648
Misura
625 x 530 x 140 mm
Descrizione generale
The BRAND® stackable tray is made from durable white polypropylene (PP), designed for versatile use in laboratory and industrial settings. Its ribbed bottom enhances stability, preventing contents from shifting during transport or storage. The reinforced rim adds strength, while the slightly sloping walls facilitate easy access and pouring. This stackable design allows for efficient storage, maximizing space when not in use. Ideal for organizing and handling various materials, this tray combines practicality with durability, making it an essential tool for any workspace.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR156648-1EA | 4061831854572 |