Documenti fondamentali
BR150922
BRAND® watch glass
PTFE, diam. 150 mm
Sinonimo/i:
Dishes, PTFE, Watch glasses
About This Item
Codice UNSPSC:
41121801
NACRES:
NB.43
Materiali
PTFE
Produttore/marchio commerciale
BRAND 150922
Diametro
150 mm
Descrizione generale
The BRAND® watch glass is made from high-quality PTFE, making it ideal for covering PTFE beakers. Its excellent chemical resistance ensures durability and reliability when handling a wide range of substances. This watch glass provides a secure cover to prevent contamination and evaporation during experiments. Its lightweight and non-reactive properties make it a valuable accessory in laboratory settings, enhancing the efficiency of various applications. This versatile tool is essential for maintaining the integrity of samples and solutions in the lab.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)