BR136315
BRAND® stirring rod
high-density polyethylene (with steel core), L × D 200 mm × 6 mm
Sinonimo/i:
PTFE, Stirring rods
About This Item
Codice UNSPSC:
41121802
NACRES:
NB.72
Prodotti consigliati
Materiali
high-density polyethylene (with steel core)
Descrizione
with steel core
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 136315
L × D
200 mm × 6 mm
Categorie correlate
Descrizione generale
The BRAND® stirring rod is designed to be bendable, allowing for customization based on specific applications. The permanent bend ensures that the rod maintains its shape during use, providing flexibility for stirring in various vessel shapes and sizes. This innovative design enhances the efficiency of mixing and blending processes, making it suitable for a wide range of laboratory tasks.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR136315-1EA | 4061831863970 |