Documenti fondamentali
BR134080
BRAND® flow indicator
for tubing I.D. 6-11 mm
Sinonimo/i:
Flow indicator, Tubing
About This Item
Codice UNSPSC:
41105108
NACRES:
NB.35
Materiali
polypropylene
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 134080
Categorie correlate
Descrizione generale
Styrene- acrylonitrile (SAN), transparent. This useful device optimizes cooling water flow during distillation. This helps to decrease operating costs by reducing water consumption. Operating pressure max. 2 bar, operating temperature for SAN max. 30 °C. For tubing of inner-Ø 6-11 mm.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)