Documenti fondamentali
BR128954
BRAND® narrow-mouth bottles, PFA
capacity 500 mL
Sinonimo/i:
Bottles, PFA, Plastics
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
PFA bottle
PFA cap
clear bottle
narrow-mouth bottle
round bottle
Giunto
Joints threaded neck (S28)
Caratteristiche
autoclavable
with cap
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 128954
Capacità
500 mL
diam. × alt.
76 mm × 190 mm
Descrizione generale
Bottles are designed for long-term storage of solvents and standards (trace analytics). PFA screw cap with a molded sealing ring for a positive seal. Autoclavable, easy to clean, withstands temperatures from -200 °C to 260 °C.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)