BR113130
BRAND® forcep with rectangular ends
PTFE, L 150 mm
Sinonimo/i:
Forceps, PTFE
About This Item
Codice UNSPSC:
41122405
NACRES:
NB.35
Prodotti consigliati
Materiali
PTFE
Produttore/marchio commerciale
BRAND 113130
Lunghezza
150 mm
Descrizione generale
The BRAND® Forceps with Rectangular Ends are specialized tools designed for handling materials in challenging laboratory environments. Made from high-quality PTFE (polytetrafluoroethylene), these forceps offer exceptional durability and performance. The PTFE construction ensures excellent resistance to a wide range of chemicals, making these forceps suitable for use in corrosive environments. Capable of withstanding high temperatures, these forceps are ideal for applications that involve heat exposure.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
BR113130-1EA | 4061831851960 |