Documenti fondamentali
BAH189620001
SP Bel-Art® Poxygrid® Desktop Microliter Pipettor Rack
to hold, Gilson Pipetman
Sinonimo/i:
Pipette Rack
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Caratteristiche
to hold Gilson Pipetman
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art H18962-0001
Lungh. × Largh. × Alt.
9 in. (229 mm) × 5 in. (127 mm) × 6.75 in. (171 mm)
Descrizione generale
Choose from 3 styles of epoxy-coated aluminum rack to hold Gilson Pipetman® pipettors.
Caratteristiche e vantaggi
  • Provides storage and easy access for up to 10 pipettors plus up to two Pipet-Aid® pipettes
  • H18962-0001 for desktop storage
  • H18962-0002 for wall mounting; has two holes for screw attachment to the wall and double faced tape (included)
  • H18962-0004 has extended base with raised lip to hold plastic tip boxes; suitable for wall mounting or for free standing on a bench
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Pipet-Aid is a registered trademark of Drummond Scientific Co.
Pipetman is a registered trademark of Gilson S.A.S. LLC
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)