BAF379001100
SP Bel-Art® Minipet Aqueous Pipettor
capacity 10 mL
Sinonimo/i:
Pipette
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art F37900-1100
Capacità
10 mL
Categorie correlate
Descrizione generale
Minipet Pipettors feature a three-way check valve for continuous, repetitive pipetting. Just place the filling line in a reservoir, push the PTFE® plunger to dispense and release to refill.
Caratteristiche e vantaggi
- Molded polypropylene handle offers comfort for extended use
Turning the knob on top sets the self-locking capacity to assure dispensing repeatability of ± 0.5%; extensions can be added to the filling line for greater reach
PTFE Minipet Pipettors offer a high level of chemical resistance; may be used with virtually any solvent or acid (except HF); fluids come in contact with only borosilicate glass and PTFE; Supplied with an all-PTFE valve (with glass check) and PTFE sinker on 45.7cm (18”) line, 2.4mm I.D.(⅛”)
Aqueous Minipet Pipettors are autoclavable and can be used with a Luer lock manifold for multi-well plate work; supplied with a chrome plated check valve, Luer lock fitting, Buna-N-seals, stainless steel spring, and Luer lock cannula, 45.7cm long (18”), 3.2mm (⅛”) I.D. silicon filling tube and PTFE sinker
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)