Merck
509655
Spritzen mit Luer-Lock-Ventil (509353)
volume 100 mL, 100MR-VLLMA-GT, needle size (not included)
About This Item
UNSPSC-Code:
41122001
Materialien
glass syringe
Kanülengröße
(not included)
Leistungsmerkmale
Pre-fitted with Luer Lock Valve
Verpackung
pkg of 1 ea
Hersteller/Markenname
SGE 009770
Volumen
100 mL
Allgemeine Beschreibung
  • SGE Syringes Fitted with Luer Lock Valve, Model: 100MR-VLLMA-GT.
  • Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger should be removed.
  • Feature an interference fit between the plunger tip and the barrel, which enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKUGTIN
5096554061833037737