Merck
Z640182
Eppendorf® epT.I.P.S. Biopur®, singles
volume 50-1000 μL
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Materialien
colorless
Sterilität
sterile
Verpackung
pack of 100 (individually wrapped)
Hersteller/Markenname
Eppendorf® 0030 010.051
Eppendorf® 022491156
Messbereich Volumen
50-1000 μL
Länge
71 mm
Maximales Volumen
1 mL
Volumen
50—1000 μL
Allgemeine Beschreibung
Biopur guaranteed sterile, pyrogen-free, RNase-free, DNA-free and ATP-free, for applications in pharmaceutical and food industries, molecular biology and cell technology.
Batch specific certificates available on the internet at http://www.eppendorf.com
Rechtliche Hinweise
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)