Merck
CLS480102
Optional heads for Corning® LSE vortex mixers
optional head for 1 microplate, 64 x 0.2 mL tubes, or 8 x 0.2 mL tube strips, 1/cs
Synonym(e):
equipment replacement accessory
About This Item
UNSPSC-Code:
41103814
NACRES:
NB.15
Verpackung
case of 1
pack of 1
Hersteller/Markenname
Corning 480102
Eignung
suitable for (for use with Corning LSE vortex mixer)
Allgemeine Beschreibung
Optional heads for the Corning LSE vortex mixers: CLS6776, CLS6777 and CLS6775
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)