Merck
CLS480101
Optional heads for Corning® LSE vortex mixers
optional head for 24 x 1.5/2.0 mL tubes, 24 x 0.5 mL tubes and 32 x 0.2 mL tubes (or 4 tube strips), 1/cs
Synonym(e):
equipment replacement accessory
About This Item
UNSPSC-Code:
41103814
NACRES:
NB.15
Verpackung
case of 1
pack of 1
Hersteller/Markenname
Corning 480101
Eignung
suitable for (for use with Corning LSE vortex mixer)
Allgemeine Beschreibung
Optional heads for the Corning LSE vortex mixers: CLS6776, CLS6777 and CLS6775
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.