CLS431644
Corning® roller bottles, not treated
bottle surface area 850 cm2, Easy grip cap, polystyrene, sterile
Synonym(e):
Corning roller bottles
About This Item
Empfohlene Produkte
Materialien
clear bottle
cylindrical bottle
high-density polyethylene cap
polystyrene
wide-mouth bottle
Sterilität
sterile
Leistungsmerkmale
cap
Verpackung
pack of 1 ea
case of 40 ea
Hersteller/Markenname
Corning 431644
Flaschenhalsdurchm.
~45 mm
Kapazität
255 mL
Durchm. × H
117 mm × 271 mm
Bindungsart
non-treated surface
Verwandte Kategorien
Leistungsmerkmale und Vorteile
- Caps are made of high-density polyethylene, whereas bottles are made of virgin polystyrene
- Flexible tubing with heat sealability and chemically resistant
- Seamless structure with printed graduations and lot number to help in product traceability
- Easy grip cap with wide knurls for easy handling
- Gamma radiation sterilized, having sterility assurance level (SAL) of 10-6
- Nonpyrogenic
- Compatible with disposable tubing set
Rechtliche Hinweise
Ähnliches Produkt
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Artikel
3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models
Global Trade Item Number
SKU | GTIN |
---|---|
CLS431644-1EA | 4061831674200 |
CLS431644-40EA | 4061831674217 |