Z742491
Generator probe for D1000 handheld homogenizer
5 x 50 mm flat bottom, for microtubes
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.61
Empfohlene Produkte
Kompatibilität mit Sterilisationsverfahren
autoclavable compatible
Leistungsmerkmale
5 x 50 mm flat bottom, for microtubes
Verpackung
pkg of 5
Hersteller/Markenname
Benchmark Scientific D1000-M5
Allgemeine Beschreibung
Generator probes for Benchmark′s handheld homogenizer, the D1000.
All the generator probes are made of stainless steel and can be sterilized by autoclaving.
All the generator probes are made of stainless steel and can be sterilized by autoclaving.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)