Merck
Z742490
Generator probe for D1000 handheld homogenizer
14 mm x 130 mm saw tooth, for 50 mL tubes up to 250 mL vessels
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.61
Kompatibilität mit Sterilisationsverfahren
autoclavable compatible
Leistungsmerkmale
14 mm x 130 mm saw tooth, for 50 mL tubes up to 250 mL vessels
Verpackung
pkg of 1
Hersteller/Markenname
Benchmark Scientific D1000-M14
Allgemeine Beschreibung
Generator probes for Benchmark′s handheld homogenizer, the D1000.
All the generator probes are made of stainless steel and can be sterilized by autoclaving.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)