Z557455
Julabo® bath spring clamps for Erlenmeyer flasks
fits 200 - 250 mL flasks
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.77
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 8970606
Allgemeine Beschreibung
The individual spring clips can be positioned in the shaking water bath using the base tray for spring clips (Z743201-1EA). A maximum of 15 spring clips for 200/250 ml Erlenmeyer flasks can be mounted on a base tray.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z557455-1EA | 4061833062197 |