Z218693
Clamps
fits flask, 1,000 mL
Synonym(e):
flask clamp, flask lab, lab flask, reaction flasks
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.43
Empfohlene Produkte
Leistungsmerkmale
fits flask 1,000 mL
Hersteller/Markenname
Ace Glass 650806
Allgemeine Beschreibung
Two-piece, black anodized aluminum clamp has three wing nuts to secure Ace reaction flask to head. Clamps also fit Pyrex reaction kettles.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z218693-1EA | 4061832348933 |