Merck
Z557471
Julabo® bath spring clamps for Erlenmeyer flasks
fits 1000 mL flasks
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.77
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 8970609
Allgemeine Beschreibung
The individual spring clips can be positioned in the shaking water bath using the base tray for spring clips (Z743201-1EA). A maximum of 4 spring clips for 1000 ml Erlenmeyer flasks can be mounted on a base tray.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z557471-1EA4061833062210