Merck
Z552909
Sigma-Aldrich
Aldrich® reaction vessel bubbler with SafetyBarb® hose connectors
joint: ST/NS 29/32
Synonym(e):
bubblers, scientific bubblers
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Verbindung
joint: ST/NS29/32
Kapazität
5-7 mL
Verwandte Kategorien
Allgemeine Beschreibung
Excellent from closing off reaction vessel from the atmosphere. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z552909-1EA4061837636073