WHA3822-090
Whatman® glass microfiber filters, Grade GF/C RTU
preweighed, pack of 100 ea
Alle Fotos(1)
About This Item
UNSPSC-Code:
40161507
NACRES:
NB.24
Empfohlene Produkte
Materialien
borosilicate glass
Leistungsmerkmale
Binder Free
preweighed
Verpackung
pack of 100 ea
Parameter
<550 °C max. temp.
105 mL/min frequency
Dicke
(260 μm)
Verwandte Kategorien
Allgemeine Beschreibung
Cytiva′s ready-to-use Whatman Grade GF/C glass microfiber filter papers offer a good flow rate and loading capacity that support total suspended solids analysis with a fine nominal particle retention of 1.2 µm.
Glass microfiber filters for total suspended solids
Whatman binder-free glass microfiber filter grade GF/C from Cytiva is widely used as a total suspended solids filter for applications testing both potable water and waste water.
The GF/C RTU line, part of the Whatman GF/C filter paper family, is pre-treated with washing, drying, cooling (in a desiccator), and weighing steps aligned to the requirements of EN 872—the European norm for testing suspended solids in water. The Cytiva quality organization certifies that these filter paper have weight loss of less than 0.017 mg/cm2 after preparation, as required by the standard. Each ready-to-use glass filter paper is placed in an individually barcoded aluminium pan that notes the filter paper weight. The barcode can be used to access a Cytiva certification of the filter weight from our website. Each box contains an additional box ID and barcode that can be used to download the weights of all 100 pans in that box.
Glass microfiber filters for total suspended solids
Whatman binder-free glass microfiber filter grade GF/C from Cytiva is widely used as a total suspended solids filter for applications testing both potable water and waste water.
The GF/C RTU line, part of the Whatman GF/C filter paper family, is pre-treated with washing, drying, cooling (in a desiccator), and weighing steps aligned to the requirements of EN 872—the European norm for testing suspended solids in water. The Cytiva quality organization certifies that these filter paper have weight loss of less than 0.017 mg/cm2 after preparation, as required by the standard. Each ready-to-use glass filter paper is placed in an individually barcoded aluminium pan that notes the filter paper weight. The barcode can be used to access a Cytiva certification of the filter weight from our website. Each box contains an additional box ID and barcode that can be used to download the weights of all 100 pans in that box.
Leistungsmerkmale und Vorteile
- Available in a range of sizes,
- GF/C grade microfiber filters are widely used for cell harvesting, liquid scintillation counting, and binding assays where more loading capacity is required.
- They are highly suitable for use in applications involving air filtration and for gravimetric analysis of volatile materials where ignition is involved.
- GF/C grade RTU microfiber filters can be used for dissolved and suspended solids and are available in packs of 100.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
WHA3822-090 |