Merck
WHA10421030
Whatman® glass microfiber grade GF 92 prefilter discs
white, inorganic binder, 200 ea, 50 mm diam
Synonym(e):
Whatman filter, Z698121, fiber filters, glass filter
About This Item
UNSPSC-Code:
40161507
NACRES:
NB.24
Produktbezeichnung
Whatman® glass microfiber filters with inorganic binder, Grade GF 92, diam. 50 mm
Materialien
borosilicate glass membrane
circles
Sterilität
non-sterile
Leistungsmerkmale
hydrophilic
Verpackung
pkg of 200 ea
Hersteller/Markenname
Whatman 10421030
Whatman Article No. 28415407 (US reference)
Durchmesser
50 mm
Allgemeine Beschreibung
This filter is used as a membrane prefilter in applications such as the determination of crop protection agent residues by GC or HPLC, in cold sludge determination of beer, in soot separation before gas analyzers and as roll filters in automatic air filtration units.
Leistungsmerkmale und Vorteile
  • Inorganic binder provides increased strength
  • Temperature resistant up to 500°C
  • Medium filtration speed
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)