SYNF740027
Synthware™ micro funnel
O.D. 27 mm
Synonym(e):
Synthware F740027, glass funnel
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.51
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
F740027
AD
27 mm
Allgemeine Beschreibung
Synthware™ micro funnel is designed for use with microscale applications. Its narrow neck allows for accurate pouring into small containers or tubes, minimizing spillage and ensuring efficient handling of samples. The glass material provides chemical resistance and visibility of the contents.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)