Merck
SYNA791924
Synthware bushing adapter
female joint: ST/NS 19/38, male joint: ST/NS 24/40
Synonym(e):
Synthware A791924, glass bushing adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
female joint: ST/NS19/38
male joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A791924
Verwandte Kategorien
Allgemeine Beschreibung
Bushing adapters allow for compact assemblies of dissimilar joints.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)