BR65243
BRAND® desiccator ′Novus′
nominal diameter × O.D. × H 250 mm × 320 mm × 425 mm, top socket joint: 24/29
Alle Fotos(2)
About This Item
UNSPSC-Code:
41104510
NACRES:
NB.43
Empfohlene Produkte
Verbindung
socket joint: top24/29
Leistungsmerkmale
lid with socket and stopcock
Hersteller/Markenname
BRAND 65243
Nenndurchmesser × AD × H
250 mm × 320 mm × 425 mm
Verwandte Kategorien
Allgemeine Beschreibung
Boro 3.3. DIN 12491. Precisely ground flat flanges. Base ring surface studded to improve stability and help prevent cracks. Interchangeable lid. Desiccators with sockets are supplied with 24/29 socket, and interchangeable Boro 3.3 stopcock.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR65243-1EA | 4061831857665 |