Merck
BR462617
BRAND® rubber conical gasket for filter funnels and filter flasks
size 28 mm
Synonym(e):
Filter, Rubber gasket
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.24
Materialien
EPDM rubber
Verpackung
pack of 10 ea
Hersteller/Markenname
BRAND 462617
Größe
28 mm
Allgemeine Beschreibung
Conical gaskets to place between filter funnel and filter flask.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)