Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R-018

Supelco

Retigabine solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18FN3O2
CAS Number:
Molecular Weight:
303.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

InChI

1S/C16H18FN3O2/c1-2-22-16(21)20-15-8-7-13(9-14(15)18)19-10-11-3-5-12(17)6-4-11/h3-9,19H,2,10,18H2,1H3,(H,20,21)

InChI key

PCOBBVZJEWWZFR-UHFFFAOYSA-N

General description

Retigabine, also known as ezogabine, is a new antiepileptic drug developed for treatment of partial seizures. The drug is marketed under the trade names Potiga in the US and Trobalt in the EU. This certified solution standard is suitable for use as starting material in calibrators and controls for LC/MS or GC/MS methods in clinical toxicology, forensic analysis, urine drug testing, pain prescription monitoring, or pharmaceutical research applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup - Solvent

Flash Point(C)

6.0 °C - closed cup - Solvent


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emma D Deeks
CNS drugs, 25(10), 887-900 (2011-09-23)
Retigabine (ezogabine in the US) opens neuronal voltage-gated potassium channels, resulting in resting membrane potential stabilization, neuronal subthreshold excitability control and anticonvulsant effects. The clinical efficacy of adjunctive oral retigabine in adults with inadequately controlled, partial-onset seizures was demonstrated in
Michele Y Splinter
Clinical therapeutics, 34(9), 1845-1856 (2012-08-25)
Ezogabine, also known as retigabine, is a recently approved anticonvulsant medication with a novel mechanism of action. It activates low-threshold voltage-gated potassium channels, leading to hyperpolarization of the membrane potential, stabilization of the resting membrane potential, and suppression of repetitive
G Blackburn-Munro et al.
CNS drug reviews, 11(1), 1-20 (2005-05-04)
Retigabine [D23129; N-(2-amino-4-(4-fluorobenzylamino)-phenyl)carbamic acid ethyl ester] is an antiepileptic drug with a recently described novel mechanism of action that involves opening of neuronal K(V)7.2-7.5 (formerly KCNQ2-5) voltage-activated K(+) channels. These channels (primarily K(V)7.2/7.3) enable generation of the M-current, a subthreshold
Piotr Czuczwar et al.
Pharmacological reports : PR, 62(2), 211-219 (2010-05-29)
Retigabine represents an antiepileptic drug possessing a completely different mechanism of action when compared to the existing classical and newer antiepileptic drugs. In the therapeutic range, retigabine enhances potassium currents, very likely via destabilization of a closed conformation or stabilization
Pingzheng Zhou et al.
Molecular pharmacology, 87(1), 31-38 (2014-10-17)
Retigabine (RTG, [ethyl N-[2-amino-4-[(4-fluorophenyl)methyl]amino] phenyl] carbamate]) is a first-in-class antiepileptic drug that acts by potentiating neuronal KCNQ potassium channels; however, it has less than optimal brain distribution. In this study, we report that P-RTG (ethyl N-[2-amino-4-((4-fluorobenzyl)(prop-2-ynyl)amino)phenyl]carbamate), an RTG derivative that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service