Merck
LC/MS Analysis of Clenbuterol Enantiomers in Plasma on Astec® CHIROBIOTIC® T after SPE using HybridSPE®-Phospholipid
LC/MS Analysis of Clenbuterol Enantiomers in Plasma on Astec® CHIROBIOTIC® T after SPE using HybridSPE®-Phospholipid application for HPLC
Materiais
Aplicação
Nº do produto
Descrição
Preços
LC/MS Analysis of Clenbuterol In Plasma on Astec® CHIROBIOTIC® T with Phospholipid Removal using HybridSPE® Phospholipid
application for SPE, application for LC-MS
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T
application for HPLC
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T2
application for HPLC
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® V
application for HPLC
Padrão
Nº do produto
Descrição
Preços
Clenbuterol hydrochloride
≥95%
coluna analítica
Nº do produto
Descrição
Preços
Coluna de HPLC quiral Astec® CHIROBIOTIC® T
5 μm particle size, L × I.D. 10 cm × 2.1 mm
tubo ou placa SPE
Nº do produto
Descrição
Preços
HybridSPE®-Phospholipid
96-well Plate, bed wt. 50 mg, volume 2 mL, pk of 1
HybridSPE®-Phospholipid
Cartridge, bed wt. 30 mg, volume 1 mL, pk of 100
CONDITIONS
column
CHIROBIOTIC T, 10 cm x 2.1 mm I.D., 5 μm particles (12018AST)
mobile phase
10 mM ammonium formate in methanol
flow rate
0.3 mL/min
column temp.
30 °C
detector
ESI(+)
sample
clenbuterol, 10 ng/mL in rat plasma (phospholipids removed by extraction with HybridSPE-Phospholipid)
Descrição
Informações legais
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany