K5142-U
Omnifit® EZ Benchmark Columns with 2 Fixed Endpieces
I.D. × L 15 mm × 500 mm
About This Item
Código UNSPSC:
23151816
NACRES:
SB.52
Produtos recomendados
fabricante/nome comercial
Omnifit 006EZ-15-50-FF
Nível de qualidade
técnica(s)
LPLC: suitable
D.I. × C
15 mm × 500 mm
Categorias relacionadas
Descrição geral
Omnifit® Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system. Two configurations are available:
- Non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.
- One fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80mm of height adjustment.
Informações legais
Omnifit is a registered trademark of Diba Industries
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
K5142-U | 4061833673003 |