Documentos Principais
22138-U
On/Off Throttling Valve
brass, 1/8 in.
About This Item
Código UNSPSC:
41112500
eCl@ss:
32035302
NACRES:
SB.54
Materiais
brass
conector
1/8 in.
Descrição geral
Nuts and ferrules included.
Throttling valve′s on and off settings determines whether or not fluid flows past the valve. It is much more efficient than the toggle on/off valve, as it provides better flow control between 0 and 100% flow and has a larger baseline pressure drop. There are many styles of throttling valve like globe valve and butterfly valve.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Process Plant Equipment: Operation, Control, and Reliability.
Holloway MD
Technology & Engineering, 401-402 (2012)