Z743572
PetriStickers™
50-square grid format
About This Item
Código UNSPSC:
41122700
NACRES:
NB.35
Produtos recomendados
embalagem
pack of 36 ea
fabricante/nome comercial
Diversified Biotech PSTK-1050
Descrição geral
Quick Reference - Petristickers correspond to wells on a microplate for fast colony identification
High Contrast - Colored grid allows colonies to stand out from the Petri dish background
Updated Design - Rectangular shape leaves ample space above and below grid for writing on the dish
Two Layouts - Reference wells numerically (1-96) or alpha numerically (A1-H12)
Ideally Sized - PetriStickers™ are 2.8 in. x 1.75 in. (71 mm x 44.5 mm), to fit most common Petri dishes
High Contrast - Colored grid allows colonies to stand out from the Petri dish background
Updated Design - Rectangular shape leaves ample space above and below grid for writing on the dish
Two Layouts - Reference wells numerically (1-96) or alpha numerically (A1-H12)
Ideally Sized - PetriStickers™ are 2.8 in. x 1.75 in. (71 mm x 44.5 mm), to fit most common Petri dishes
Informações legais
PetriStickers is a trademark of Diversified Biotech
p3xFLAG is a trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)