Documentos Principais
Z709867
Sartorius pipette tips
volume range 0.2-10 μL, standard, racked, sterile
About This Item
Código UNSPSC:
41121600
NACRES:
NB.01
volume:
10 μL
Materiais
colorless
esterilidade
sterile
Características
DNase free
RNase free
barrier: no
endotoxin free
embalagem
pack of 96 ea (racked)
fabricante/nome comercial
Sartorius 790011
volume máximo
10 μL
faixa de volume
0.2-10 μL
volume
10 μL
Descrição geral
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Outras notas
Browse all pipette tips
Escolha uma das versões mais recentes:
Certificados de análise (COA)