Z376892
Nichiryo digital hand-held dispenser
dispensing volume 2.0-10.0 mL
About This Item
Código UNSPSC:
41121500
NACRES:
NB.51
Produtos recomendados
Características
electronic: no
single-channel
embalagem
pack of 1 ea
fabricante/nome comercial
Nichiryo 00-21DG-10
dispensing volume measuring range
2.0-10.0 mL
volume
2 × 10 mL
Categorias relacionadas
Descrição geral
This light weight dispenser allows for continuous dispensing of solutions. Glass cylinder and plunger are protected from breakage by plastic housings. All parts which come in direct contact with the solution are autoclavable. Provides accuracy of ±1% and precision of <0.2%.
Escolha uma das versões mais recentes:
Certificados de análise (COA)