Documentos Principais
Z374385
Sigma-Aldrich
Scienceware® slide box
for 100 microscope slides, Hinged lid
About This Item
Código UNSPSC:
41123302
NACRES:
MA.01
Características
Holds 100 x slides 76x26 mm
embalagem
pack of 1 ea
fabricante/nome comercial
Bel-Art H44577-0000
Descrição geral
Supreme defense for standard 25 x 75mm (1" x 3") microscope slides
  • Heavy-walled, moisture- and insect-proof.
  • Slim, durable design
  • Easily identify slides with included numbered index on base and cover
  • Box walls are rugged and won′t bend, split, or chip.
Informações legais
Scienceware is a registered trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 8
1 of 8
BRAND® slide box Holds 50 x slides 76x26 mm
BR475900
BRAND® slide box
Premium Slide box 100 Place, cork lining, blue
HS15994A
Premium Slide box
Premium slide box 100 place, cork lining, gray plastic
HS15994F
Premium slide box
BRAND® slide box 25 slides, 76 x 26 mm
BR475800
BRAND® slide box
Premium slide box 100 place, cork lining, white plastic
HS15994E
Premium slide box
Slide box 25 place, red
HS15990B
Slide box
Premium Slide box 25 Place, cork lining, blue
HS15989A
Premium Slide box