Documentos Principais
Z329835
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 250 mL, label, Ethanol, with venting valve (metal ball)
About This Item
Código UNSPSC:
41103209
NACRES:
NB.17
Materiais
low-density polyethylene bottle
orange closure
round bottle
translucent bottle
wide-mouth bottle
Características
label Ethanol
with cap
with venting valve (metal ball)
embalagem
pack of 5 ea
fabricante/nome comercial
BRAND 144022
capacidade
250 mL
Descrição geral
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Outras notas
BR144022 (India)
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
acessório
Nº do produto
Descrição
Preços
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Global Trade Item Number
SKUGTIN
Z329835-5EA4061831842197