OH30456394
Ohaus® Navigator® Portable Balance
Model NVT12000, weighing capacity 12000 g, resolution: 1 g, AC/DC input 230 V, universal plug set
About This Item
Código UNSPSC:
41111502
NACRES:
NB.71
Produtos recomendados
Materiais
metal weighing plate
Características
CE compliant
embalagem
pack of 1 ea
fabricante/nome comercial
Ohaus® 30456394
capacidade de pesagem faixa de medição
12000 g, resolution: 1 g
Tacho de pesagem: L × C
17.4 cm × 23 cm
Descrição geral
OHAUS raises the bar in price-performance ratio with the newly-designed Navigator series which is perfect for general weighing applications in laboratory, industrial and education settings. Ruggedly constructed and multifunctional, the Navigator′s a powerful balance that can handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity, and easy operation.
Aplicação
Weighing, Parts Counting, Percent Weighing, Checkweighing
Características e benefícios
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Informações legais
Navigator is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
OH30456394-1EA | 4065266751185 |