GBL666581
Grace Bio-Labs Press-To-Seal silicone isolator, red silicone sheet
thickness 0.8 mm
Sinônimo(s):
Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier
About This Item
Produtos recomendados
Materiais
red silicon
Características
(Adhesive on both sides)
embalagem
pack of 5 ea
fabricante/nome comercial
Grace Bio-Labs 666581
C × L
180 mm × 130 mm
espessura
0.8 mm
Categorias relacionadas
Descrição geral
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
GBL666581-5EA | 4061832400952 |