Documentos Principais
EP5910754000
Adapter for Eppendorf® S-4x Universal Large Rotor
for 16x75-100 mm round-bottom tubes, pkg of 2 ea
About This Item
Código UNSPSC:
41103911
NACRES:
NB.78
Características
Holds 7.5 - 12 mL tubes
for 16x75-100 mm round-bottom tubes
embalagem
pkg of 2 ea
fabricante/nome comercial
Eppendorf® 5910754000
técnica(s)
centrifugation: suitable
compatibilidade
for use with Universal bucket
Descrição geral
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Informações legais
Eppendorf is a registered trademark of Eppendorf AG