Documentos Principais
EP5895120008
Rotor for Eppendorf® 5920R Centrifuges
swing-bucket rotor, S-4x750, aerosol-tight, with 4 × 750 mL round buckets (open), with Eppendorf QuickLock® lid
About This Item
Código UNSPSC:
41103909
NACRES:
NB.82
Características
to hold x 0.2 mL to 750 mL tubes and flasks
to hold x plates
aerosol-tight
swing-bucket rotor
with 4 × 750 mL round buckets (open)
with Eppendorf QuickLock® lid
embalagem
pack of 1 ea
fabricante/nome comercial
Eppendorf® 5895120008 (Global)
Parâmetros
4,800 × g max. RCF
técnica(s)
centrifugation: suitable
compatibilidade
for use with centrifuge models 5920R
Descrição geral
The large Centrifuge 5920 R is a benchtop centrifuge that combines exceptional capacity with a compact and ergonomic design. It has similar dimensions to refrigerated competitor models in the 3-liter segment, but has an outstanding capacity of up to 4 x 1,000 mL or 52 x 50 mL tubes. Optimize high-throughput centrifugation and make your daily work easier.
Informações legais
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf QuickLock is a registered trademark of Eppendorf AG
Escolha uma das versões mais recentes:
Certificados de análise (COA)