EP5353040156
Eppendorf® Tube Holder
25/50 mL
About This Item
Código UNSPSC:
41121703
NACRES:
NB.11
Produtos recomendados
Características
25/50 mL
embalagem
case of 1 ea
fabricante/nome comercial
Eppendorf® 5353040156
Categorias relacionadas
Descrição geral
- High-speed mixer (3,000 rpm) for fast and efficient mixing of small volumes in all common vessel formats up to 50 mL
- 2DMix-Control technology (planar mixing orbit without vertical movement) for controlled mixing and reproducible results in seconds
- Anti-spill technology for controlled mixing movements without lid wetting and cross-contamination
- 5 different tube holders (0.5 mL, 1.5/2.0 mL, 5/15 mL, 30/50 mL, PCR 96-well plates) for high vessel flexibility
- Integrated ergonomic vortex function for various vessel formats for distinct sample mixing
- Minimal hand/arm vibration transfer during vortexing (certificate available) for ergonomic operation
- 5 soft keys with pre-defined standard mixing parameters for easy usage
- Very stable - remains stationary even at maximum mixing speed
- Whisper quiet operation to benefit the work environment
Informações legais
Eppendorf is a registered trademark of Eppendorf AG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
EP5353040156-1EA | 4061841436485 |