Key Documents
EP0030120973
Eppendorf® micropestles
pkg of 10 ea
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
embalagem
pkg of 10 ea
fabricante/nome comercial
Eppendorf® 0030120973
Descrição geral
  • Eppendorf® micropestles are accessories for sample handling.
  • They are used for resuspending pellets and comprise of two sides for optimal fit in 1.5 mL and 2.0 mL vessels.
  • Autoclavable (121 °C, 20 min).
Informações legais
Eppendorf is a registered trademark of Eppendorf AG