Documentos Principais
BR31576
BRAND® BLAUBRAND® measuring cylinder with serialized numbers
capacity 500 mL
About This Item
Código UNSPSC:
41121800
embalagem
pkg of 12 ea
fabricante/nome comercial
BRAND 27452
Altura
115 mm
capacidade
500 mL
Descrição geral
Measuring cylinders tall form. Class A DE-M marking to DIN EN ISO 4788. Made from DURAN(R), calibrated to contain (TC, In). With spout and hexagonal glass base. Ring marks at major graduations. High contrast blue enamel graduations and inscription
Informações legais
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)