BAH445830000
Slide Tray Cabinet
Holds 12 Microscope Slide Trays
About This Item
Código UNSPSC:
41122800
NACRES:
NB.22
Produtos recomendados
embalagem
pack of 1 ea
fabricante/nome comercial
Bel-Art H44583-0000
tamanho
17 × 36 × 17 cm
Categorias relacionadas
Descrição geral
This compact cabinet organizes and stores microscope slides. The small footprint of the cabinet means it can be placed on the benchtop, making the slides simply accessible and close to the point of use.
- Compact cabinet to organize and store microscope slides
- Stores 240 slides in 12 microscope trays (not included)
- Dust free storage
- Polypropylene and polystyrene construction
- Transparent vertically sliding acrylic door with door knob
Escolha uma das versões mais recentes:
Certificados de análise (COA)