Z743109
Julabo® flow through immersion cooler
FD 200, temp. range 10 °C (30 °C), AC/DC input 230 V AC, Schuko plug
About This Item
Código UNSPSC:
41103706
NACRES:
NB.77
Produtos recomendados
embalagem
pack of 1 ea
fabricante/nome comercial
FD 200
Julabo 9655825
Parâmetros
10 °C temp. range (30 °C)
Descrição geral
JULABO flow-through coolers are the perfect complement to open bath circulators, immersion circulators and bridge mounted circulators extending their temperature range to below ambient temperature. They offer a small footpri nt, have a low energy requirement and represent an economic solution preventing tap water usage for cooling. Instruments are designed for integration into an existing cooling loop (e.g. consisting of open bath circulator wit h pump).
Your advantages
Your advantages
- For applications near ambient temperatures
- Integrated freezing protection
- Preventing the use of precious tap water for cooling
- Compact design, small footprint
- Ease of operation
- Low energy requirement
- Time saving - Comfortable and simple operation for setpoint adjustment
- Precise Temperature Control - PID Temperature control with set control parameters, temperature stability ±0.02...±0.2 °C
Informações legais
Julabo is a registered trademark of Julabo USA, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z743109-1EA |