Documentos Principais
Z685291
Platinum crucible, with reinforced rim
capacity 30 mL
About This Item
Código UNSPSC:
41122200
NACRES:
NB.43
fabricante/nome comercial
Birmingham metals
capacidade
30 mL
Descrição geral
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. These platinum crucibles can be cleaned by boiling in a suitable solvent, for the removal of substances which have alloyed with the surface, scour with alumina powder. Having a reinforced rim means that the rim of the crucible is thicker than the walls to make it stronger; this is where the crucible can be gripped if tongs are used during the handling process.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 8
1 of 8
Platinum crucible standard, capacity 15 mL
Z562041
Platinum crucible
Platinum crucible lid fits 50 mL crucible
Z562122
Platinum crucible lid
Platinum evaporating dish capacity 100 mL
Z562203
Platinum evaporating dish
Platinum crucible micro, capacity 5 mL
Z562238
Platinum crucible
Platinum crucible standard, capacity 10 mL
Z562025
Platinum crucible
Global Trade Item Number
SKUGTIN
Z685291-1EA4061833513484