Documentos Principais
Z675741
Eyela aspirator vacuum pump
AC/DC input 230 V AC, UK plug
About This Item
Código UNSPSC:
41100000
NACRES:
NB.79
fabricante/nome comercial
Eyela 210538 UK CORD
Descrição geral
Safely aspirates corrosive and volatile solvents without cold or chemical traps. Vapors are condensed and diluted in an internal 19 L PP reservoir for easy disposal.
Maximum pump speed of 19 L/min at 20°C achieves a vacuum of 23.3 mbar (2338 Pa) at a water temperature of 20°C. Pumping method is pressed recirculation by water-jet pump. Use with rotary evaporators, distillation or filtration apparatus, and electrophoresis gel driers.
Escolha uma das versões mais recentes:
Certificados de análise (COA)