Z568694
Aldrich® combustion apparatus flask, with stopper and pinch clamp
capacity 1000 mL
About This Item
Código UNSPSC:
41121804
NACRES:
NB.43
Produtos recomendados
Materiais
borosilicate glass
conical
single-neck
esterilidade
non-sterile
Características
closure type Stopper
embalagem
pack of 1
fabricante/nome comercial
Sigma-Aldrich
capacidade
1000 mL
Categorias relacionadas
Descrição geral
Heavy-wall borosilicate glass conical flask with 35/25 ball joint stopper. Use with a detachable platinum sample carrier suspended from the stopper hook. Pinch clamp holds stopper in place during combustion.
Order platinum carrier and sample wrappers separately.
Based on a design by C.L. Ogg, R.B. Kelly, and J.A. Connelly. See Microchemical Techniques, N.D. Cheronis, Ed. Interscience, 1962, pp 427-433.
Order platinum carrier and sample wrappers separately.
Based on a design by C.L. Ogg, R.B. Kelly, and J.A. Connelly. See Microchemical Techniques, N.D. Cheronis, Ed. Interscience, 1962, pp 427-433.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z568694-1EA | 4061832965307 |