Z562637
Aldrich® lecture-bottle station
for general service and storage
About This Item
Código UNSPSC:
41121800
NACRES:
NB.51
Produtos recomendados
entrada da CGA
180
Descrição geral
Dispensing lecture-bottle gases has never been safer or simpler using these portable lecture-bottle stations. The premium quality components housed within a station are matched to the application, fully assembled, leak-tested, and ready for use.
Just connect the station to the lecture-bottle, process, and a source of inert gas, if using a station with a purge valve, and you are ready to go.
Safe and Simple:
Station Specifications:
1. Heavy-gauge steel cabinet with durable, red enamel finish, table-top or wall mountable
2. Quality, panel-mounted regulators and purge valves
3. Braided, 316L stainless steel hose with CGA-180 remote connection to the lecture-bottle
4. Support for two lecture-bottles, one in use and a spare
5. Clearly labeled gauges, gas connections, and valve functions
6. Instructions for Use label on front of cabinet
7. Fully assembled, leak-tested and ready for use
Just connect the station to the lecture-bottle, process, and a source of inert gas, if using a station with a purge valve, and you are ready to go.
Safe and Simple:
- Gas dispensing and storage in one compact, portable system
- Keeps lecture-bottles upright and stable, eliminates tip-over
- Reduces equipment damage and chance of personal injury
- Choice of four models matched to the application
- Online Lecture-Bottle Equipment to Gases Selection Guide
Station Specifications:
1. Heavy-gauge steel cabinet with durable, red enamel finish, table-top or wall mountable
2. Quality, panel-mounted regulators and purge valves
3. Braided, 316L stainless steel hose with CGA-180 remote connection to the lecture-bottle
4. Support for two lecture-bottles, one in use and a spare
5. Clearly labeled gauges, gas connections, and valve functions
6. Instructions for Use label on front of cabinet
7. Fully assembled, leak-tested and ready for use
This station provides storage capacity for three additional lecture-bottles, tools, and supplies for use with Sigma-Aldrich Lecture-Bottle Stations. Table top or wall-mountable. Hinged cover over storage space keeps supplies clean and secure.
Ligação
For more information on station set-up, see the Lecture Bottle Technical Bulletin
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
produto relacionado
Nº do produto
Descrição
Preços
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z562637-1EA | 4061826764077 |