Z557579
Aldrich® pear-shaped splash-guard adapter
capacity 100 mL, female joint: ST/NS 24/40
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
Produtos recomendados
Junta
female joint: ST/NS24/40
male joint ST/NS14/20
descrição
anti-climb
capacidade
100 mL
Descrição geral
Application: for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping. Tapered sidewalls eliminate hold up common to round style adapters and permits continuous washing with condensed solvent.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)